Learn More
Eisco™ Dicot Leaf , Cross Section
Catalog No. S30387
Change view
Click to view available options
Quantity:
1
Catalog No. Quantity
S30387 1
1 options
Catalog No. S30387 Supplier Eisco™ Supplier No. BS18246
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Prepared microscope slide of a cross section of a dicot leaf. Showing general structures. Great choice for biology classrooms to explore structure function relationships as per NGSS standards.
  • pre-cleaned glass slide with ground edges
  • size 1x3 in. (25x77 mm)
  • professionally stained samples for best visualization
  • great choice for biology classrooms to explore structure-function relationships as per NGSS standards
  • showing characteristic structures
Specifications
Material Glass
Type Prepared Microscope Slide
For Use With (Application) Microscopy
Quantity 1
Specimen Type Dicot Leaf
Size 1 x 3 in.
Slide Type Prepared Slide