missing translation for 'onlineSavingsMsg'
Learn More

Eisco™ Excretory System Ureter, Cross Section

Catalog No. S30390
Change view
Click to view available options
Quantity:
1
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S30390 1
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S30390 Supplier Eisco™ Supplier No. BS18249

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Prepared microscope of a cross section of mammalian ureter, a part of excretory system. Stained to show characteristic tissues, including transitional epithelium. Transitional epithelium is comprised of two or more layers of cuboidal or columnar cells. Great for biology classrooms.

  • pre-cleaned glass slide with ground edges
  • size 1x3 in. (25x77 mm)
  • professionally stained samples for best visualization
  • great choice for biology classrooms to explore structure-function relationships as per NGSS standards
  • showing characteristic tissues, including transitional epithelium

Specifications

Description Shows characteristic tissues, including transitional epithelium, which is comprised of 2+ layers of cuboidal or columnar cells
Classification Excretory System
Material Glass
Type Prepared Microscope Slide
For Use With (Application) Microscopy
Quantity 1
Specimen Type Mammalian Ureter
Size 1 x 3 in.
Slide Type Prepared Slide