missing translation for 'onlineSavingsMsg'
Learn More

Eisco™ Spinal Cord (Human), Cervical Cross Section

Catalog No. S30355
Change view
Click to view available options
Grade:
9 to 12
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Grade
S30355 9 to 12
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S30355 Supplier Eisco™ Supplier No. BS18208

Prepared microscope slide of a cross section of a human cervical spinal cord. Stained for better visualization of characteristic structures. Great choice for biology classrooms studying nervous system.

  • Pre-cleaned glass slide with ground edges
  • Size 1x3 in. (25x77 mm)
  • Professionally stained samples for best visualization
  • Great choice for biology classrooms to explore structure-function relationships as per NGSSs standards
  • Showing characteristic structures

Specifications

Description Cervical Cross Section
Classification Cell Biology
Color Clear
Material Glass
Mounting Type Cross Section
Type Prepared Microscope Slide
For Use With (Application) Microscopy
Grade 9 to 12
Quantity 1 Ea.
Species Human
Specimen Type Spinal Cord
Size 1 x 3 in.
Slide Type Prepared Slide
Show More Show Less