missing translation for 'onlineSavingsMsg'
Learn More

Eisco™ Penis, Cross Section, H&E Stain

Catalog No. S30354
Change view
Click to view available options
Quantity:
1 Ea.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S30354 1 Ea.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S30354 Supplier Eisco™ Supplier No. BS18207
Only null left

Prepared microscope slide of a cross section of a mamallian penis. Stained with hematoxylin and eosin for better visualization of characteristic structures such as urethra and corpora cavernosa. Great choice for biology classrooms to explore structure function relationships as per NGSS standards.

  • Pre-cleaned glass slide with ground edges
  • Size 1x3 in. (25x77 mm)
  • Professionally stained samples for best visualization
  • Great choice for biology classrooms to explore structure-function relationships as per NGSSs standards
  • Showing characteristic structures such as urethra and corpora cavernosa

Specifications

Description Cross Section shows characteristic structures such as urethra and corpora cavernosa
Classification Reproductive System
Material Glass
Type Prepared Microscope Slide
For Use With (Application) Microscopy
Quantity 1 Ea.
Specimen Type Penis
Stain Type H and E stain
Size 1 x 3 in.
Slide Type Prepared Slide