missing translation for 'onlineSavingsMsg'
Learn More

Eisco™ Artery, Vein and Nerve (Human ), Cross Section

Catalog No. S30349
Change view
Click to view available options
Quantity:
1 Ea.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S30349 1 Ea.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S30349 Supplier Eisco™ Supplier No. BS18202

Prepared microscope slide of a cross section of human artery, vein and nerve. Stained for beter visualization of characteristic structures and tissues. Side by side placement of artery, vein, and nerve allows for easy comparison of the three tissues on one slide.

  • Pre-cleaned glass slide with ground edges
  • Size 1x3 in. (25x77 mm)
  • Professionally stained samples for best visualization
  • Great choice for biology classrooms to explore structure-function relationships as per NGSSs standards
  • Showing side by side artery, vein and nerve and characteristic structures of these tissues

Specifications

Description Cross Section shows side by side artery, vein and nerve in their characteristic structures
Material Glass
Type Prepared Microscope Slide
For Use With (Application) Microscopy
Quantity 1 Ea.
Species Human
Specimen Type Artery, Vein, and Nerve
Size 1 x 3 in.
Slide Type Prepared Slide