missing translation for 'onlineSavingsMsg'
Learn More

Eisco™ Retina and Tapetum (Mammal), H&E Stain

Catalog No. S30359
Change view
Click to view available options
Quantity:
1 Ea.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S30359 1 Ea.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S30359 Supplier Eisco™ Supplier No. BS18214

Prepared microscope slide of mammalian retina and tapetum cross section. Stained with Hematoxylin and Eosin for better visualization of eye structures, including the optic nerve entrance. Great choice for biology classrooms to explore structure function relationships as per NGSS standards.

  • Pre-cleaned glass slide with ground edges
  • Size 1x3 in. (25x77 mm)
  • Professionally stained samples for best visualization
  • Great choice for biology classrooms to explore structure-function relationships as per NGSSs standards
  • Showing eye structures, including the optic nerve entrance

Specifications

Description Shows eye structures, including the optic nerve entrance
Material Glass
Type Prepared Microscope Slide
For Use With (Application) Microscopy
Quantity 1 Ea.
Species Mammal
Specimen Type Retina and Tapetum
Stain Type H and E stain
Size 1 x 3 in.
Slide Type Prepared Slide