missing translation for 'onlineSavingsMsg'
Learn More

Eisco™ Hair Follicle

Catalog No. S30674
Change view
Click to view available options
Grade:
9 to 12
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Grade
S30674 9 to 12
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S30674 Supplier Eisco™ Supplier No. BS18122

Prepared microscope slide of hair follicle. Showing hair follicles, epidermis, and other skin structures. Great choice for biology classrooms to explore structure function relationships as per NGSS standards.

  • Pre-cleaned glass slide with ground edges
  • Size 1x3 in. (25x77 mm)
  • Professionally stained samples for best visualization
  • Great choice for biology classrooms to explore structure-function relationships as per NGSSs standards
  • Showing characteristic structures including ovules and embryo sacs

Specifications

Description Shows characteristic structures, including primary follicles and epidermis
Classification Microbiology
Color Clear
Material Glass
Type Prepared Microscope Slide
For Use With (Application) Microscopy
Grade 9 to 12
Quantity 1 Ea.
Specimen Type Hair Follicle
Size 1 x 3 in.
Slide Type Prepared Slide
Show More Show Less