missing translation for 'onlineSavingsMsg'
Learn More

Eisco™ Respiratory System - Pneumonia (Human)

Catalog No. S30627
Change view
Click to view available options
Grade:
9 to 12
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Grade
S30627 9 to 12
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S30627 Supplier Eisco™ Supplier No. BS18278

Prepared microscope slide of human lungs affected with pneumonia. Stained for better visualization of characteristic pathological signs of pneumonia such as thickened alveolar walls and infiltrating immune cells. Great choice for biology classrooms.

  • Pre-cleaned glass slide with ground edges
  • Size 1x3 in. (25x77 mm)
  • Professionally stained samples for best visualization
  • Great choice for biology classrooms to explore structure-function relationships as per NGSSs standards
  • Showing characteristic pathological signs of pneumonia such as thickened alveolar walls and infiltrating i mmune cells

Specifications

Description Shows characteristic pathological signs of pneumonia infected lungs such as thickened alveolar walls and infiltrating immune cells
Classification Respiratory System
Color Clear
Material Glass
Type Prepared Microscope Slide
For Use With (Application) Microscopy
Grade 9 to 12
Quantity 1 Ea.
Species Human
Specimen Type Pneumonia Infected Lungs
Size 1 x 3 in.
Slide Type Prepared Slide
Show More Show Less