missing translation for 'onlineSavingsMsg'
Learn More

Eisco™ Female Reproductive System - Immature Ovary

Catalog No. S30367
Change view
Click to view available options
Grade:
9 to 12
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Grade
S30367 9 to 12
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S30367 Supplier Eisco™ Supplier No. BS18222
Only null left

Prepared microscope slide of mammalian immature ovary. Ovaries are a part of female reproductive system. Stained for better visualization of characteristic structures, including primary follicles. Great choice for biology classrooms to explore structure function relationships as per NGSS standards.

  • Pre-cleaned glass slide with ground edges
  • Size 1x3 in. (25x77 mm)
  • Professionally stained samples for best visualization
  • Great choice for biology classrooms to explore structure-function relationships as per NGSSs standards
  • Showing characteristic structures, including primary follicles

Specifications

Description Shows characteristic structures including ovules and embryo sacs
Classification Female Reproductive System
Color Clear
Material Glass
Mounting Type Cross Section
Type Prepared Microscope Slide
For Use With (Application) Microscopy
Grade 9 to 12
Quantity 1 Ea.
Species Human
Specimen Type Immature Ovary
Size 1 x 3 in.
Slide Type Prepared Slide
Show More Show Less