missing translation for 'onlineSavingsMsg'
Learn More

Eisco™ Amphiuma Liver Section

Catalog No. S30388
Change view
Click to view available options
Grade:
9 to 12
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Grade
S30388 9 to 12
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S30388 Supplier Eisco™ Supplier No. BS18247

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Prepared microscope slide of a section of amphiuma liver. Amphiuma is a genus of aquatic salamanders. Showing characteristic structures of typical animal cells. Great choice for biology classrooms to explore structure function relationships as per NGSS standards.

  • pre-cleaned glass slide with ground edges
  • size 1x3 in. (25x77 mm)
  • professionally stained samples for best visualization
  • great choice for biology classrooms to explore structure-function relationships as per NGSS standards
  • showing characteristic structures of typical animal cells

Specifications

Classification Aquatic Salamander
Material Glass
Type Prepared Microscope Slide
For Use With (Application) Microscopy
Grade 9 to 12
Quantity 1
Specimen Type Amphiuma Liver
Size 1 x 3 in.
Slide Type Prepared Slide