missing translation for 'onlineSavingsMsg'
Learn More

Eisco™ Jejunum Section (Mammal)

Catalog No. S30384
Change view
Click to view available options
Quantity:
1
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S30384 1
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S30384 Supplier Eisco™ Supplier No. BS18243

Prepared microscope slide of mammalian jejunum. Jejunum is a part of small intestine. Stained for better visualization of characteristic layers of small intestines. Great choice for biology classrooms studying digestive sysem.

  • pre-cleaned glass slide with ground edges
  • size 1x3 in. (25x77 mm)
  • professionally stained samples for best visualization
  • great choice for biology classrooms to explore structure-function relationships as per NGSS standards
  • showing characteristic layers of mammalian small intestines

Specifications

Classification Small Intestine
Material Glass
Type Prepared Microscope Slide
For Use With (Application) Microscopy
Quantity 1
Specimen Type Mammal Jejunum
Size 1 x 3 in.
Slide Type Prepared Slide