missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Eisco™ Female Reproductive System - Vagina
Description
- pre-cleaned glass slide with ground edges
- size 1x3 in. (25x77 mm)
- professionally stained samples for best visualization
- great choice for biology classrooms to explore structure-function relationships as per NGSS standards
- showing characteristic tissues and structures
Specifications
Specifications
| Classification | Reproductive System |
| Material | Glass |
| Type | Prepared Microscope Slide |
| For Use With (Application) | Microscopy |
| Quantity | 1 |
| Specimen Type | Female Vagina |
| Size | 1 x 3 in. |
| Slide Type | Prepared Slide |