missing translation for 'onlineSavingsMsg'
Learn More

G-Biosciences Prep Desalting Column

Catalog No. s03098
Encompass
Change view
Click to view available options
Quantity:
5 x 15 mL
5 x 30 mL
1 x 150 mL
5 x 150 mL
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S03098 5 x 30 mL
S03096 5 x 150 mL
S03097 1 x 150 mL
S03099 5 x 15 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. S03098 Supplier G-Biosciences Supplier No. 7862015
Only null left

Prep Desalting columns are ready to use prepacked columns for separation of low molecular weight substances from high molecular weight substances based on size exclusion chromatography.

  • Prep Desalting columns are mostly used for removal of salt or buffer exchange before or after different chromatographic steps.
  • Prep Desalting columns can be used for separation of proteins based on their sizes.
  • G‐Biosciences offer different sizes of desalting columns.
  • Prep Desalting columns are made of biocompatible polypropylene, which does not interact with biomolecules.
  • The column has a stopper at the inlet and one top plug, one end cap, two filter discs.

Specifications

Product Type Desalting Column
Format Gravity Flow Column
Column Type Size-exclusion, Proprietary Resin
MWCO 5.0 kDa
Product Line Pierce
Disposable Reusable
Quantity 5 x 30 mL
Sample Volume (Metric) 30 mL