Learn More
Eisco™ Premium Bunsen Burner
Includes flame stabilizer, air mix adjustment, and gas flow adjustment for complete control in the laboratory
Supplier:  Eisco™ CH0094NG
 View more versions of this product
Catalog No. S27507
Only null left
Explore available promotions
Description
Description
  • Flame stabilizing burner tip for increased safety
  • Includes an air port with collar to adjust the flow of air
  • Gas flow is controlled by a needle valve fitted above the base
  • Burner comes equipped with a serrated inlet arm that accepts 6-8 mm tubing
  • Aluminum mixing tube is mounted on a circular, die cast zinc alloy base
Specifications
Specifications
Bunsen Burner
All
165.1 mm
Natural Gas
Fits 6 to 8 mm tubing
6.5 in.
Aluminum, Die Cast Zinc Alloy
Product Suggestions
Product Suggestions