missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Pregnenolone Acetate 99.0+%, TCI Americaâ„¢
$62.37 - $183.30
Chemical Identifiers
CAS | 1778-02-5 |
---|---|
Molecular Formula | C23H34O3 |
Molecular Weight (g/mol) | 358.522 |
MDL Number | MFCD00003627 |
InChI Key | CRRKVZVYZQXICQ-BODHGXFRSA-N |
Synonym | 1s,5s,10s,11s,14s,15s,2r-14-acetyl-2,15-dimethyltetracyclo 8.7.0.0<2,7>.0<11 ,15> heptadec-7-en-5-yl acetate, 11as-1-acetyl-9a,11a-dimethyl-1h,2h,3h,3ah,3bh,4h,6h,7h,8h,9h,9bh,10h,11h-cyclopenta a phenanthren-7-yl acetate, 13s-17-ethanoyl-10,13-dimethyl-2,3,4,7,8,9,11,12,14,15,16,17-dodecahydro-1h-cyclopenta a phenanthren-3-yl ethanoate, acetic acid 13s-17-acetyl-10,13-dimethyl-2,3,4,7,8,9,11,12,14,15,16,17-dodecahydro-1h-cyclopenta a phenanthren-3-yl ester |
PubChem CID | 18530247 |
IUPAC Name | [(13S)-17-acetyl-10,13-dimethyl-2,3,4,7,8,9,11,12,14,15,16,17-dodecahydro-1H-cyclopenta[a]phenanthren-3-yl] acetate |
SMILES | CC(=O)C1CCC2C1(CCC3C2CC=C4C3(CCC(C4)OC(=O)C)C)C |
Chemical Identifiers
1778-02-5 | |
358.522 | |
CRRKVZVYZQXICQ-BODHGXFRSA-N | |
18530247 | |
CC(=O)C1CCC2C1(CCC3C2CC=C4C3(CCC(C4)OC(=O)C)C)C |
C23H34O3 | |
MFCD00003627 | |
1s,5s,10s,11s,14s,15s,2r-14-acetyl-2,15-dimethyltetracyclo 8.7.0.0<2,7>.0<11 ,15> heptadec-7-en-5-yl acetate, 11as-1-acetyl-9a,11a-dimethyl-1h,2h,3h,3ah,3bh,4h,6h,7h,8h,9h,9bh,10h,11h-cyclopenta a phenanthren-7-yl acetate, 13s-17-ethanoyl-10,13-dimethyl-2,3,4,7,8,9,11,12,14,15,16,17-dodecahydro-1h-cyclopenta a phenanthren-3-yl ethanoate, acetic acid 13s-17-acetyl-10,13-dimethyl-2,3,4,7,8,9,11,12,14,15,16,17-dodecahydro-1h-cyclopenta a phenanthren-3-yl ester | |
[(13S)-17-acetyl-10,13-dimethyl-2,3,4,7,8,9,11,12,14,15,16,17-dodecahydro-1H-cyclopenta[a]phenanthren-3-yl] acetate |
Specifications
1778-02-5 | |
White-Yellow | |
MFCD00003627 | |
1s,5s,10s,11s,14s,15s,2r-14-acetyl-2,15-dimethyltetracyclo 8.7.0.0<2,7>.0<11 ,15> heptadec-7-en-5-yl acetate, 11as-1-acetyl-9a,11a-dimethyl-1h,2h,3h,3ah,3bh,4h,6h,7h,8h,9h,9bh,10h,11h-cyclopenta a phenanthren-7-yl acetate, 13s-17-ethanoyl-10,13-dimethyl-2,3,4,7,8,9,11,12,14,15,16,17-dodecahydro-1h-cyclopenta a phenanthren-3-yl ethanoate, acetic acid 13s-17-acetyl-10,13-dimethyl-2,3,4,7,8,9,11,12,14,15,16,17-dodecahydro-1h-cyclopenta a phenanthren-3-yl ester | |
CC(=O)C1CCC2C1(CCC3C2CC=C4C3(CCC(C4)OC(=O)C)C)C | |
358.522 | |
358.52 | |
Crystalline Powder |
151°C | |
C23H34O3 | |
1 g | |
CRRKVZVYZQXICQ-BODHGXFRSA-N | |
[(13S)-17-acetyl-10,13-dimethyl-2,3,4,7,8,9,11,12,14,15,16,17-dodecahydro-1H-cyclopenta[a]phenanthren-3-yl] acetate | |
18530247 | |
≥99.0% (HPLC) | |
Pregnenolone Acetate |
Safety and Handling
EINECSNumber : (4)-1300
TSCA : No