missing translation for 'onlineSavingsMsg'
Learn More

Pregnenolone Acetate 99.0+%, TCI America™

Catalog No. p047710g
Change view
Click to view available options
Quantity:
1 g
10 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P047710G 10 g
P04771G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P047710G Supplier TCI America Supplier No. P047710G
Only null left

Chemical Identifiers

CAS 1778-02-5
Molecular Formula C23H34O3
Molecular Weight (g/mol) 358.522
MDL Number MFCD00003627
InChI Key CRRKVZVYZQXICQ-BODHGXFRSA-N
Synonym 1s,5s,10s,11s,14s,15s,2r-14-acetyl-2,15-dimethyltetracyclo 8.7.0.0<2,7>.0<11 ,15> heptadec-7-en-5-yl acetate, 11as-1-acetyl-9a,11a-dimethyl-1h,2h,3h,3ah,3bh,4h,6h,7h,8h,9h,9bh,10h,11h-cyclopenta a phenanthren-7-yl acetate, 13s-17-ethanoyl-10,13-dimethyl-2,3,4,7,8,9,11,12,14,15,16,17-dodecahydro-1h-cyclopenta a phenanthren-3-yl ethanoate, acetic acid 13s-17-acetyl-10,13-dimethyl-2,3,4,7,8,9,11,12,14,15,16,17-dodecahydro-1h-cyclopenta a phenanthren-3-yl ester
PubChem CID 18530247
IUPAC Name [(13S)-17-acetyl-10,13-dimethyl-2,3,4,7,8,9,11,12,14,15,16,17-dodecahydro-1H-cyclopenta[a]phenanthren-3-yl] acetate
SMILES CC(=O)C1CCC2C1(CCC3C2CC=C4C3(CCC(C4)OC(=O)C)C)C

Specifications

Melting Point 151°C
Color White-Yellow
Quantity 10 g
Formula Weight 358.52
Percent Purity ≥99.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material Pregnenolone Acetate
EINECSNumber (4)-1300
TSCA No