missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Prednisolone 98.0+%, TCI America™
$63.38 - $667.33
Chemical Identifiers
| CAS | 50-24-8 |
|---|---|
| Molecular Formula | C21H28O5 |
| Molecular Weight (g/mol) | 360.45 |
| MDL Number | MFCD00003649 |
| InChI Key | OIGNJSKKLXVSLS-VWUMJDOOSA-N |
| Synonym | prednisolone, metacortandralone, hydroretrocortine, predonine, deltacortril, meticortelone, deltahydrocortisone, prenolone, hydroretrocortin, delta-cortef |
| PubChem CID | 5755 |
| ChEBI | CHEBI:8378 |
| IUPAC Name | (8S,9S,10R,11S,13S,14S,17R)-11,17-dihydroxy-17-(2-hydroxyacetyl)-10,13-dimethyl-7,8,9,11,12,14,15,16-octahydro-6H-cyclopenta[a]phenanthren-3-one |
| SMILES | CC12CC(C3C(C1CCC2(C(=O)CO)O)CCC4=CC(=O)C=CC34C)O |
Chemical Identifiers
| 50-24-8 | |
| 360.45 | |
| OIGNJSKKLXVSLS-VWUMJDOOSA-N | |
| 5755 | |
| (8S,9S,10R,11S,13S,14S,17R)-11,17-dihydroxy-17-(2-hydroxyacetyl)-10,13-dimethyl-7,8,9,11,12,14,15,16-octahydro-6H-cyclopenta[a]phenanthren-3-one |
| C21H28O5 | |
| MFCD00003649 | |
| prednisolone, metacortandralone, hydroretrocortine, predonine, deltacortril, meticortelone, deltahydrocortisone, prenolone, hydroretrocortin, delta-cortef | |
| CHEBI:8378 | |
| CC12CC(C3C(C1CCC2(C(=O)CO)O)CCC4=CC(=O)C=CC34C)O |
Specifications
| 50-24-8 | |
| White | |
| MFCD00003649 | |
| prednisolone, metacortandralone, hydroretrocortine, predonine, deltacortril, meticortelone, deltahydrocortisone, prenolone, hydroretrocortin, delta-cortef | |
| CC12CC(C3C(C1CCC2(C(=O)CO)O)CCC4=CC(=O)C=CC34C)O | |
| 360.45 | |
| CHEBI:8378 | |
| ≥98.0% (HPLC) | |
| Prednisolone |
| 235°C | |
| C21H28O5 | |
| 1 g | |
| OIGNJSKKLXVSLS-VWUMJDOOSA-N | |
| (8S,9S,10R,11S,13S,14S,17R)-11,17-dihydroxy-17-(2-hydroxyacetyl)-10,13-dimethyl-7,8,9,11,12,14,15,16-octahydro-6H-cyclopenta[a]phenanthren-3-one | |
| 5755 | |
| 360.45 | |
| Crystalline Powder |
Safety and Handling
RTECSNumber : TU4152000
TSCA : Yes