missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Precoated TLC and PLC Glass Plates

Average particle size is 15μm with a 5-20μm distribution

Supplier:  MilliporeSigma™ 1.13894.0001

 View more versions of this product

Catalog No. M1138940001


Only null left

Description

Description

  • Choose from several media to match your separation requirements: silica gel 60 (with and without F254 fluorescent indicator), chemically modified silica gel (reversed phases RP-2, RP-8 and RP-18), aluminum oxide 60 with F254 indicator, cellulose, and cellulose PEI
  • 60Å silica gel is suitable for all common gel separations including weak to strong polar analytes
  • Silica can also be used for applications requiring charring procedures
  • Aluminum oxide or alumina plates are ideal for separating hormones, alkaloids and antibiotics
  • For partition chromatography, select cellulose plates; and for separation of weak anion exchangers, including peptide and amino acids, use cellulose PEI plates

Certifications

ISO 9001:2000

Specifications

Chemical Identifiers

Glass

Specifications

Glass
20/Pk.
Cleaning up Synthetic Reaction Mixtures, Natural Products, Plant Extracts and Biotech Products
Silica 60 (0.5 mm)
Silica Gel PLC Plate
20 x 20 cm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions