missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MilliporeSigma™ Precoated TLC and PLC Glass Plates
Average particle size is 15μm with a 5-20μm distribution
Supplier: MilliporeSigma™ 1.05738.0001
Description
- Choose from several media to match your separation requirements: silica gel 60 (with and without F254 fluorescent indicator), chemically modified silica gel (reversed phases RP-2, RP-8 and RP-18), aluminum oxide 60 with F254 indicator, cellulose, and cellulose PEI
- 60Å silica gel is suitable for all common gel separations including weak to strong polar analytes
- Silica can also be used for applications requiring charring procedures
- Aluminum oxide or alumina plates are ideal for separating hormones, alkaloids and antibiotics
- For partition chromatography, select cellulose plates; and for separation of weak anion exchangers, including peptide and amino acids, use cellulose PEI plates
Certifications
ISO 9001:2000
Chemical Identifiers
| Glass |
Specifications
| Glass | |
| Kieselguhr F254 | |
| 25/Pk. | |
| 254 nm | |
| Kieselguhr F254 | |
| 20 cm |
| ISO 9001:2000 | |
| 60 Å | |
| Kieselguhr TLC Plate | |
| Separation of Polar and Moderately Polar Substances, Flavonoids and Other Phenolic Substances | |
| 20 x 20 cm |