Learn More
MilliporeSigma™ Precoated HPTLC Glass Plates
Smaller size and distribution of HPTLC media yields improved resolution, shorter analysis times, and higher detection sensitivity
$615.27 - $1879.58
| Catalog Number | Mfr. No. | Dimensions (L x W) | Quantity | Sorbent | Price | Quantity | |||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Dimensions (L x W) | Quantity | Sorbent | Price | Quantity | |||
|
M1056420001
|
MilliporeSigma™
1056420001 |
20 x 10 cm | 50/Pk. | Silica Gel 60 F254 |
Each for $1,202.68
|
|
|||
|
M1056290001
|
MilliporeSigma™
1056290001 |
10 x 10 cm | 100/Pk. | Silica Gel 60 F254 |
Each for $1,879.58
|
|
|||
|
M1056330001
|
MilliporeSigma™
1056330001 |
10 x 10 cm | 100/Pk. | Silica Gel 60 |
Each for $1,550.68
|
|
|||
|
M1056410001
|
MilliporeSigma™
1056410001 |
20 x 10 cm | 50/Pk. | Silica Gel 60 |
Each for $1,188.79
|
|
|||
|
M1156960001
|
MilliporeSigma™
1156960001 |
20 x 10 cm | 25/Pk. | Silica Gel 60 F254 s |
Each for $1,375.24
|
|
|||
|
M1056480001
|
MilliporeSigma™
1056480001 |
20 x 10 cm | 50/Pk. | Silica Gel 60 Premium |
Each for $615.94
|
|
|||
|
M1056440001
|
MilliporeSigma™
1056440001 |
5 x 5 cm | 100/Pk. | Silica Gel 60 Prescored |
Each for $1,286.96
|
|
|||
|
M1056310001
|
MilliporeSigma™
1056310001 |
10 x 10 cm | 25/Pk. | Silica Gel 60 |
Each for $615.27
|
|
|||
|
M1056280001
|
MilliporeSigma™
1056280001 |
10 x 10 cm | 25/Pk. | Silica Gel 60 F254 |
Each for $677.13
|
|
|||
Description
- Average particle size is 5μm with a 4-7μm distribution
- Untreated, chemically modified, and reversed-phase silica gel with a pore size of 60Å
For all common gel separations, including weak to strong polar analytes, for applications requiring charring procedures
Certifications
ISO 9001:2000
Specifications
| Silica HPTLC Plate | |
| 200 μm |