Learn More
MilliporeSigma™ Supelco™ Precision Sampling Syringe Needles
Catalog No. 111020934
Change view
Click to view available options
Needle Point Style:
Cone with Side Port
For Use With (Equipment):
VICI Series A
VICI Series A-2, C, D
Volume (Metric):
100 to 10,000 μL
25 to 10,000 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Needle Point Style For Use With (Equipment) Volume (Metric)
111020934 Cone with Side Port VICI Series A-2, C, D 25 to 10,000 μL
111011559 Cone with Side Port VICI Series A 100 to 10,000 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 111020934 Supplier MilliporeSigma™ Supelco™ Supplier No. SGMA22289
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Ideal for collection, trace analysis, and sample concentration of environmental contaminants
Specifications
Material Stainless Steel (Tip)
Description A-2, C, D
Diameter (English) Inner Needle 0.012 in.
Diameter (English) Outer Needle 0.029 in.
Length (English) Needle 2 in.
Product Type Syringe Needle
Needle Point Number 5
Quantity 3 Pk.
Volume (Metric) 25 to 10,000 μL
Needle Point Style Cone with Side Port
For Use With (Equipment) VICI Series A-2, C, D
Show More Show Less