missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ PREB Polyclonal Antibody

Goat Polyclonal Antibody
Supplier: Invitrogen™ PA5149027
Description
Reconstitute at 0.2 mg/mL in sterile PBS.
Prolactin (PRL) expression in the pituitary is limited to specific cells. Pit-1 is a pituitary specific transcription factor that plays an important role in PRL expression, both in mature organism and during development. The PRL promoter contains numerous Pit-1 binding sites and these sites have been implicated in both basal level and kinase-mediated gene expression. The most proximal of these binding sites, termed 1P, has been shown to direct a response to numerous signals, such as cAMP and various G-proteins. A novel protein, termed PREB (prolactin regulatory element binding protein), has been recently identified that regulates PRL gene expression through the 1P site, though it contains no discernable DNA-binding motif. Recent studies suggest that PREB is encoded by a single-copy gene in both mice and humans and exhibits nuclear accumulation in pituitary cells. Evidence suggests that PREB is a novel transcription factor that assists in PRL expression whether alone, or in concert with Pit-1.Chemical Identifiers
| PREB | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Polyclonal | |
| Lyophilized | |
| IgG | |
| Human | |
| Preb | |
| C85705; mammalian guanine nucleotide exchange factor mSec12; PREB; prolactin regulatory binding-element protein; prolactin regulatory element binding; prolactin regulatory element binding protein; prolactin regulatory element-binding protein; Sec12 | |
| Preb | |
| Primary | |
| Antigen affinity chromatography |
| 0.2 mg/mL | |
| Western Blot | |
| Unconjugated | |
| Goat | |
| RUO | |
| PBS with trehalose and no preservative | |
| Q9HCU5 | |
| 10113 | |
| E. coli-derived recombinant human PREB, Met1-Ser388. | |
| Antibody |
Specifications
| PREB | |
| Polyclonal | |
| Unconjugated | |
| Preb | |
| C85705; mammalian guanine nucleotide exchange factor mSec12; PREB; prolactin regulatory binding-element protein; prolactin regulatory element binding; prolactin regulatory element binding protein; prolactin regulatory element-binding protein; Sec12 | |
| Goat | |
| Antigen affinity chromatography | |
| RUO | |
| 10113 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Lyophilized |
| Western Blot | |
| 0.2 mg/mL | |
| PBS with trehalose and no preservative | |
| Q9HCU5 | |
| Preb | |
| E. coli-derived recombinant human PREB, Met1-Ser388. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG |