missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ PRDM11 Polyclonal Antibody

Description
Immunogen sequence: QVDFWFCESC QEYFVDECPN HGPPVFVSDT PVPVGIPDRA ALTIPQGMEV VKDTSGESDV RCVNEVIPKG HIFGPYEGQI STQD Highest antigen sequence indentity to the following orthologs: Mouse - 96%, Rat - 93%.
PRDM11 (PR/SET Domain 11) is a Protein Coding gene. Gene Ontology (GO) annotations related to this gene include nucleic acid binding and methyltransferase activity. May be involved in transcription regulation.
Specifications
Specifications
| Antigen | PRDM11 |
| Applications | Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Concentration | 0.05 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 |
| Gene | PRDM11 |
| Gene Accession No. | Q9NQV5 |
| Gene Alias | 8030443D09Rik; AA163972; Gm707; PFM8; PR domain 11; PR domain containing 11; PR domain-containing protein 11; PRDM11; PR-domain containing protein 11; RGD1305673 |
| Gene Symbols | PRDM11 |
| Show More |