missing translation for 'onlineSavingsMsg'
Learn More

Prazosin hydrochloride

An alpha adrenoceptor antagonist | CAS: 19237-84-4 | C19H22ClN5O4 | 419.87 g/mol

Supplier:  Thermo Scientific Chemicals J6171203

 View more versions of this product

Catalog No. AAJ6171203


Only null left
Explore available promotions

Description

Description

Prazosin hydrochloride is an inhibitor of PDE. Also offered in a free base form Prazosin

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Prazosin hydrochloride is an inhibitor of PDE. Also offered in a free base form Prazosin

Solubility
Soluble in dimethyl sulfoxide, ethanol, water, methanol.

Notes
Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
Specifications

Specifications

Prazosin hydrochloride
285°C
C19H22ClN5O4
1 g
14,7717
Soluble in dimethyl sulfoxide,ethanol,water,methanol.
[H+].[Cl-].COC1=C(OC)C=C2C(N)=NC(=NC2=C1)N1CCN(CC1)C(=O)C1=CC=CO1
419.87
CHEBI:8365
Powder
19237-84-4
White
MFCD00058177
4303561
prazosin hydrochloride, prazosin hcl, minipress, vasoflex, peripress, furazosin hydrochloride, deprazolin, hypovase, hypovasole, pratsiol
WFXFYZULCQKPIP-UHFFFAOYSA-N
hydrogen 2-[4-(furan-2-carbonyl)piperazin-1-yl]-6,7-dimethoxyquinazolin-4-amine chloride
68546
419.86

Chemical Identifiers

19237-84-4
419.87
WFXFYZULCQKPIP-UHFFFAOYSA-N
68546
hydrogen 2-[4-(furan-2-carbonyl)piperazin-1-yl]-6,7-dimethoxyquinazolin-4-amine chloride
C19H22ClN5O4
MFCD00058177
prazosin hydrochloride, prazosin hcl, minipress, vasoflex, peripress, furazosin hydrochloride, deprazolin, hypovase, hypovasole, pratsiol
CHEBI:8365
[H+].[Cl-].COC1=C(OC)C=C2C(N)=NC(=NC2=C1)N1CCN(CC1)C(=O)C1=CC=CO1
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H361-H302-H315-H319-H335
Suspected of damaging fertility or the unborn child.
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P201-P202-P261-P264b-P270-P271-P280i-P281-P301+P312-P302+P352-P304+P340-P305+P351+P338-P308+P313-P330-P332+P313-P362-P501c

H302-H315-H319-H335-H361

EINECSNumber : 242-903-4

RTECSNumber : VA1350000

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only