accessibility menu, dialog, popup

UserName

CAS RN 19237-84-4

IUPAC Name: hydrogen 2-[4-(furan-2-carbonyl)piperazin-1-yl]-6,7-dimethoxyquinazolin-4-amine chloride
Synonyms: prazosin hydrochloride prazosin hcl prazosin clorhidrato
Molecular Weight (g/mol): 419.87
Molecular Formula: C19H22ClN5O4
InChi Key: WFXFYZULCQKPIP-UHFFFAOYSA-N
SMILES: [H+].[Cl-].COC1=C(OC)C=C2C(N)=NC(=NC2=C1)N1CCN(CC1)C(=O)C1=CC=CO1
Physical Form 
  • (2)
  • (2)
Quantity 
  • (2)
  • (1)
  • (1)
Percent Purity 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)