missing translation for 'onlineSavingsMsg'
Learn More

Prazosin Hydrochloride 98.0+%, TCI America™

Catalog No. p-7134857
Change view
Click to view available options
Quantity:
100 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P0938100MG 100 mg
P09381G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P0938100MG Supplier TCI America Supplier No. P0938100MG

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 19237-84-4
Molecular Formula C19H22ClN5O4
Molecular Weight (g/mol) 419.87
MDL Number MFCD00058177
InChI Key WFXFYZULCQKPIP-UHFFFAOYSA-N
Synonym prazosin hydrochloride, prazosin hcl, minipress, vasoflex, peripress, furazosin hydrochloride, deprazolin, hypovase, hypovasole, pratsiol
PubChem CID 68546
ChEBI CHEBI:8365
IUPAC Name hydrogen 2-[4-(furan-2-carbonyl)piperazin-1-yl]-6,7-dimethoxyquinazolin-4-amine chloride
SMILES [H+].[Cl-].COC1=C(OC)C=C2C(N)=NC(=NC2=C1)N1CCN(CC1)C(=O)C1=CC=CO1

Specifications

Melting Point 285°C
Color White-Yellow
Quantity 100 mg
Formula Weight 419.87
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Prazosin Hydrochloride
RTECSNumber VA1350000
TSCA No