missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Specifications
| Praziquantel | |
| 140°C | |
| C19H24N2O2 | |
| 25 g | |
| FSVJFNAIGNNGKK-UHFFFAOYSA-N | |
| 2-(cyclohexanecarbonyl)-3,6,7,11b-tetrahydro-1H-pyrazino[2,1-a]isoquinolin-4-one | |
| 4891 | |
| ≥98.0% (HPLC,N) |
| 55268-74-1 | |
| White | |
| MFCD00058531 | |
| praziquantel, droncit, biltricide, pyquiton, cesol, azinox, embay 8440, cutter tape tabs, prasiquantel, praziquantelum | |
| C1CCC(CC1)C(=O)N2CC3C4=CC=CC=C4CCN3C(=O)C2 | |
| 312.413 | |
| 312.41 | |
| Crystalline Powder |
Chemical Identifiers
| 55268-74-1 | |
| 312.413 | |
| FSVJFNAIGNNGKK-UHFFFAOYSA-N | |
| 4891 | |
| C1CCC(CC1)C(=O)N2CC3C4=CC=CC=C4CCN3C(=O)C2 |
| C19H24N2O2 | |
| MFCD00058531 | |
| praziquantel, droncit, biltricide, pyquiton, cesol, azinox, embay 8440, cutter tape tabs, prasiquantel, praziquantelum | |
| 2-(cyclohexanecarbonyl)-3,6,7,11b-tetrahydro-1H-pyrazino[2,1-a]isoquinolin-4-one |
Safety and Handling
RTECSNumber : UQ4150000
TSCA : No
Recommended Storage : Freezer