missing translation for 'onlineSavingsMsg'
Learn More

Praziquantel, 98%

Catalog No. p-7122676
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ6618806 5 g
AAJ6618814 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAJ6618806 Supplier Thermo Scientific Chemicals Supplier No. J6618806
Only null left

Used against schistosome and many cestode infestations. It is used to study voltage-gated Ca2+ channels and is a potential small molecule neurogenic. | CAS: 55268-74-1 | C19H24N2O2 | 312.413 g/mol

Praziquantel is a potent anthelmintic used against schistosome and many cestode infestations. It is used to study voltage-gated Ca2+ channels and is a potential small molecule neurogenic.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Praziquantel is a potent anthelmintic used against schistosome and many cestode infestations. It is used to study voltage-gated Ca2+ channels and is a potential small molecule neurogenic.

Solubility
Freely soluble in ethanol or dichloromethane. Slightly soluble in water

Notes
Research Use only: Not intended for animal or human diagnostic or therapeutic use.

Chemical Identifiers

CAS 55268-74-1
Molecular Formula C19H24N2O2
Molecular Weight (g/mol) 312.413
MDL Number MFCD00058531
InChI Key FSVJFNAIGNNGKK-UHFFFAOYSA-N
Synonym praziquantel, droncit, biltricide, pyquiton, cesol, azinox, embay 8440, cutter tape tabs, prasiquantel, praziquantelum
PubChem CID 4891
IUPAC Name 2-(cyclohexanecarbonyl)-3,6,7,11b-tetrahydro-1H-pyrazino[2,1-a]isoquinolin-4-one
SMILES C1CCC(CC1)C(=O)N2CC3C4=CC=CC=C4CCN3C(=O)C2

Specifications

Melting Point 136°C to 138°C
Color White
Boiling Point 1377°C
Flash Point >110°C (230°F)
Quantity 5 g
Beilstein 761557
Merck Index 14,7716
Solubility Information Freely soluble in ethanol or dichloromethane; Slightly soluble in water
Formula Weight 312.41
Percent Purity 98%
Physical Form Powder
Chemical Name or Material Praziquantel
Show More Show Less
EINECSNumber 259-559-6
RTECSNumber UQ4150000
TSCA No
storageNote1 2° to 8°C
Recommended Storage Keep cold

RUO – Research Use Only