missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Pravastatin Sodium 98.0+%, TCI America™
Supplier: TCI America P1796500MG
Specifications
| Pravastatin Sodium | |
| White-Yellow | |
| Crystalline Powder | |
| C23H35NaO7 | |
| pravastatin sodium, mevalotin, elisor, pravaselect, lipostat, selektine, pravachol, bristacol, prareduct, pravacol | |
| [Na+].CC[C@H](C)C(=O)O[C@H]1C[C@H](O)C=C2C=C[C@H](C)[C@H](CC[C@@H](O)C[C@@H](O)CC([O-])=O)[C@@H]12 | |
| 446.52 | |
| CHEBI:8361 | |
| ≥98.0% (HPLC,T) |
| 173°C | |
| 81131-70-6 | |
| 500 mg | |
| MFCD00887601,MFCD00887601 | |
| VWBQYTRBTXKKOG-IYNICTALSA-M | |
| sodium (3R,5R)-7-[(1S,2S,6S,8S,8aR)-6-hydroxy-2-methyl-8-{[(2S)-2-methylbutanoyl]oxy}-1,2,6,7,8,8a-hexahydronaphthalen-1-yl]-3,5-dihydroxyheptanoate | |
| 16759173 | |
| 446.52 |
Chemical Identifiers
| 81131-70-6 | |
| 446.52 | |
| VWBQYTRBTXKKOG-IYNICTALSA-M | |
| 16759173 | |
| sodium (3R,5R)-7-[(1S,2S,6S,8S,8aR)-6-hydroxy-2-methyl-8-{[(2S)-2-methylbutanoyl]oxy}-1,2,6,7,8,8a-hexahydronaphthalen-1-yl]-3,5-dihydroxyheptanoate |
| C23H35NaO7 | |
| MFCD00887601,MFCD00887601 | |
| pravastatin sodium, mevalotin, elisor, pravaselect, lipostat, selektine, pravachol, bristacol, prareduct, pravacol | |
| CHEBI:8361 | |
| [Na+].CC[C@H](C)C(=O)O[C@H]1C[C@H](O)C=C2C=C[C@H](C)[C@H](CC[C@@H](O)C[C@@H](O)CC([O-])=O)[C@@H]12 |
Safety and Handling
RTECSNumber : QJ7185000
TSCA : No
Recommended Storage : Refrigerator