CAS RN 81131-70-6
CAS RN 81131-70-6
IUPAC Name:
sodium (3R,5R)-7-[(1S,2S,6S,8S,8aR)-6-hydroxy-2-methyl-8-{[(2S)-2-methylbutanoyl]oxy}-1,2,6,7,8,8a-hexahydronaphthalen-1-yl]-3,5-dihydroxyheptanoate
Synonyms:
pravastatin sodium
Molecular Weight (g/mol):
446.52
Molecular Formula:
C23H35NaO7
InChi Key:
VWBQYTRBTXKKOG-IYNICTALSA-M
SMILES:
[Na+].CC[C@H](C)C(=O)O[C@H]1C[C@H](O)C=C2C=C[C@H](C)[C@H](CC[C@@H](O)C[C@@H](O)CC([O-])=O)[C@@H]12
1
–
1
of
1
results
1
Pravastatin Sodium 98.0+%, TCI America™
CAS: 81131-70-6 Molecular Formula: C23H35NaO7 Molecular Weight (g/mol): 446.52 MDL Number: MFCD00887601,MFCD00887601 InChI Key: VWBQYTRBTXKKOG-IYNICTALSA-M Synonym: pravastatin sodium,mevalotin,elisor,pravaselect,lipostat,selektine,pravachol,bristacol,prareduct,pravacol PubChem CID: 16759173 ChEBI: CHEBI:8361 IUPAC Name: sodium (3R,5R)-7-[(1S,2S,6S,8S,8aR)-6-hydroxy-2-methyl-8-{[(2S)-2-methylbutanoyl]oxy}-1,2,6,7,8,8a-hexahydronaphthalen-1-yl]-3,5-dihydroxyheptanoate SMILES: [Na+].CC[C@H](C)C(=O)O[C@H]1C[C@H](O)C=C2C=C[C@H](C)[C@H](CC[C@@H](O)C[C@@H](O)CC([O-])=O)[C@@H]12