missing translation for 'onlineSavingsMsg'
Learn More

Pravastatin Sodium 98.0+%, TCI America™

Supplier:  TCI America P1796100MG

 View more versions of this product

Catalog No. P1796100MG


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

Pravastatin Sodium
White-Yellow
Crystalline Powder
C23H35NaO7
pravastatin sodium, mevalotin, elisor, pravaselect, lipostat, selektine, pravachol, bristacol, prareduct, pravacol
[Na+].CC[C@H](C)C(=O)O[C@H]1C[C@H](O)C=C2C=C[C@H](C)[C@H](CC[C@@H](O)C[C@@H](O)CC([O-])=O)[C@@H]12
446.52
CHEBI:8361
≥98.0% (HPLC,T)
173°C
81131-70-6
100 mg
MFCD00887601,MFCD00887601
VWBQYTRBTXKKOG-IYNICTALSA-M
sodium (3R,5R)-7-[(1S,2S,6S,8S,8aR)-6-hydroxy-2-methyl-8-{[(2S)-2-methylbutanoyl]oxy}-1,2,6,7,8,8a-hexahydronaphthalen-1-yl]-3,5-dihydroxyheptanoate
16759173
446.52

Chemical Identifiers

81131-70-6
446.52
VWBQYTRBTXKKOG-IYNICTALSA-M
16759173
sodium (3R,5R)-7-[(1S,2S,6S,8S,8aR)-6-hydroxy-2-methyl-8-{[(2S)-2-methylbutanoyl]oxy}-1,2,6,7,8,8a-hexahydronaphthalen-1-yl]-3,5-dihydroxyheptanoate
C23H35NaO7
MFCD00887601,MFCD00887601
pravastatin sodium, mevalotin, elisor, pravaselect, lipostat, selektine, pravachol, bristacol, prareduct, pravacol
CHEBI:8361
[Na+].CC[C@H](C)C(=O)O[C@H]1C[C@H](O)C=C2C=C[C@H](C)[C@H](CC[C@@H](O)C[C@@H](O)CC([O-])=O)[C@@H]12
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

RTECSNumber : QJ7185000

TSCA : No

Recommended Storage : Refrigerator

SDS
Documents

Documents

Promotions

Promotions