missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Prasugrel, 98%

Chemical Identifiers

CAS 150322-43-3
Molecular Formula C20H20FNO3S
Molecular Weight (g/mol) 373.44
InChI Key DTGLZDAWLRGWQN-UHFFFAOYSA-N
Synonym prasugrel, effient, efient, prasugrel effient, prasita, 5-2-cyclopropyl-1-2-fluorophenyl-2-oxoethyl-4,5,6,7-tetrahydrothieno 3,2-c pyridin-2-yl acetate, 2-2-acetyloxy-6,7-dihydrothieno 3,2-c pyridin-5 4h-yl-1-cyclopropyl-2-2-fluorophenyl ethanone, effient tn, ethanone, 2-2-acetyloxy-6,7-dihydrothieno 3,2-c pyridin-5 4h-yl-1-cyclopropyl-2-2-fluorophenyl
PubChem CID 6918456
ChEBI CHEBI:87723
IUPAC Name [5-[2-cyclopropyl-1-(2-fluorophenyl)-2-oxoethyl]-6,7-dihydro-4H-thieno[3,2-c]pyridin-2-yl] acetate
SMILES CC(=O)OC1=CC2=C(S1)CCN(C2)C(C3=CC=CC=C3F)C(=O)C4CC4
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AC461482500
View Documents
Thermo Scientific™
461482500
250mg N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AC461480010
View Documents
Thermo Scientific™
461480010
1g N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Chemical Identifiers

150322-43-3
373.44
prasugrel, effient, efient, prasugrel effient, prasita, 5-2-cyclopropyl-1-2-fluorophenyl-2-oxoethyl-4,5,6,7-tetrahydrothieno 3,2-c pyridin-2-yl acetate, 2-2-acetyloxy-6,7-dihydrothieno 3,2-c pyridin-5 4h-yl-1-cyclopropyl-2-2-fluorophenyl ethanone, effient tn, ethanone, 2-2-acetyloxy-6,7-dihydrothieno 3,2-c pyridin-5 4h-yl-1-cyclopropyl-2-2-fluorophenyl
CHEBI:87723
CC(=O)OC1=CC2=C(S1)CCN(C2)C(C3=CC=CC=C3F)C(=O)C4CC4
C20H20FNO3S
DTGLZDAWLRGWQN-UHFFFAOYSA-N
6918456
[5-[2-cyclopropyl-1-(2-fluorophenyl)-2-oxoethyl]-6,7-dihydro-4H-thieno[3,2-c]pyridin-2-yl] acetate

Specifications

150322-43-3
97.5% min. (HPLC)
250mg
DTGLZDAWLRGWQN-UHFFFAOYSA-N
[5-[2-cyclopropyl-1-(2-fluorophenyl)-2-oxoethyl]-6,7-dihydro-4H-thieno[3,2-c]pyridin-2-yl] acetate
6918456
373.44
Prasugrel
Authentic
C20H20FNO3S
prasugrel, effient, efient, prasugrel effient, prasita, 5-2-cyclopropyl-1-2-fluorophenyl-2-oxoethyl-4,5,6,7-tetrahydrothieno 3,2-c pyridin-2-yl acetate, 2-2-acetyloxy-6,7-dihydrothieno 3,2-c pyridin-5 4h-yl-1-cyclopropyl-2-2-fluorophenyl ethanone, effient tn, ethanone, 2-2-acetyloxy-6,7-dihydrothieno 3,2-c pyridin-5 4h-yl-1-cyclopropyl-2-2-fluorophenyl
CC(=O)OC1=CC2=C(S1)CCN(C2)C(C3=CC=CC=C3F)C(=O)C4CC4
373.44
CHEBI:87723
98%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Harmful if swallowed.

GHS P Statement IF SWALLOWED: rinse mouth. Do NOT induce vomiting. Call a POISON CENTER or doctor/physician if you feel unwell. Wash face, hands and any exposed skin thoroughly after handling.

GHS Signal Word: Warning

SDS
Documents

Documents

RUO – Research Use Only