missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Prasugrel, 98%

Supplier:  Thermo Scientific™ 461480010

 View more versions of this product

Catalog No. AC461480010

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Specifications

Prasugrel
Authentic
Glass Bottle
1g
DTGLZDAWLRGWQN-UHFFFAOYSA-N
[5-[2-cyclopropyl-1-(2-fluorophenyl)-2-oxoethyl]-6,7-dihydro-4H-thieno[3,2-c]pyridin-2-yl] acetate
6918456
373.44
150322-43-3
97.5% min. (HPLC)
C20H20FNO3S
prasugrel, effient, efient, prasugrel effient, prasita, 5-2-cyclopropyl-1-2-fluorophenyl-2-oxoethyl-4,5,6,7-tetrahydrothieno 3,2-c pyridin-2-yl acetate, 2-2-acetyloxy-6,7-dihydrothieno 3,2-c pyridin-5 4h-yl-1-cyclopropyl-2-2-fluorophenyl ethanone, effient tn, ethanone, 2-2-acetyloxy-6,7-dihydrothieno 3,2-c pyridin-5 4h-yl-1-cyclopropyl-2-2-fluorophenyl
CC(=O)OC1=CC2=C(S1)CCN(C2)C(C3=CC=CC=C3F)C(=O)C4CC4
373.44
CHEBI:87723
98%

Chemical Identifiers

150322-43-3
373.44
prasugrel, effient, efient, prasugrel effient, prasita, 5-2-cyclopropyl-1-2-fluorophenyl-2-oxoethyl-4,5,6,7-tetrahydrothieno 3,2-c pyridin-2-yl acetate, 2-2-acetyloxy-6,7-dihydrothieno 3,2-c pyridin-5 4h-yl-1-cyclopropyl-2-2-fluorophenyl ethanone, effient tn, ethanone, 2-2-acetyloxy-6,7-dihydrothieno 3,2-c pyridin-5 4h-yl-1-cyclopropyl-2-2-fluorophenyl
CHEBI:87723
CC(=O)OC1=CC2=C(S1)CCN(C2)C(C3=CC=CC=C3F)C(=O)C4CC4
C20H20FNO3S
DTGLZDAWLRGWQN-UHFFFAOYSA-N
6918456
[5-[2-cyclopropyl-1-(2-fluorophenyl)-2-oxoethyl]-6,7-dihydro-4H-thieno[3,2-c]pyridin-2-yl] acetate
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
Harmful if swallowed.

GHS P Statement IF SWALLOWED: rinse mouth. Do NOT induce vomiting. Call a POISON CENTER or doctor/physician if you feel unwell. Wash face, hands and any exposed skin thoroughly after handling.

GHS Signal Word: Warning

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only