missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Praseodymium Standard For ICP, 1000μg/mL in 2% HNO3, Assurance Grade, SPEX CertiPrep™

ISO/IEC Guide 34 Certified, NIST-Traceable Certified Reference Material

$227.71 - $412.06

Chemical Identifiers

CAS 7440-10-0
Molecular Formula Pr
Molecular Weight (g/mol) 140.908
InChI Key PUDIUYLPXJFUGB-UHFFFAOYSA-N
PubChem CID 23942
ChEBI CHEBI:49828
IUPAC Name praseodymium
SMILES [Pr]
View More Specs

Includes: NIST Traceable Certificate of Analysis accredited to ISO/IEC 17025 and ISO/IEC Guide 34, Safety Data Sheet

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
PLPR2-2Y
View Documents Promotion Details
Spex Certiprep Llc
PLPR22Y
125mL Triple-leached LDPE bottle
Each for $227.71
 
PLPR2-2X
View Documents Promotion Details
Spex Certiprep Llc
PLPR22X
500mL Triple-leached LDPE bottle
Each for $412.06
 
Description

Description

Inorganic Standard manufactured for AA, ICP, ICP/MS, IC, XRF, and other analytical instrumentation

AnalyteConcentration
Pr1000μg/mL

Certifications

Accredited by A2LA for ISO/IEC 17025 and ISO/IEC Guide 34 and certified by UL-DQS to ISO 9001

Compliance

GHS/OSHA Complaint

Specifications

Chemical Identifiers

7440-10-0
140.908
23942
praseodymium
Pr
PUDIUYLPXJFUGB-UHFFFAOYSA-N
CHEBI:49828
[Pr]

Specifications

Matrix    Concentration    Cat. No.
2-5% HNO3    1000μg/mL    PLPR2-2Y, -2X    10,000    PLPR2-3Y, -3X
0.1,2.0,97.9
Pr
[Pr]
140.908
CHEBI:49828
Triple-leached LDPE bottle
2% HNO3
AA, ICP, ICP/MS, IC, XRF, and other analytical instrumentation
Praseodymium Standard
7440-10-0,7697-37-2,7732-18-5
0.1,2.0,97.9
PUDIUYLPXJFUGB-UHFFFAOYSA-N
praseodymium
23942
Plasma
Assurance Certificate of Analysis reports up to 39 elements at ppb levels.
125mL
Up to 68 elements are scanned with found values for ICP and ICP-MS standards. The found value of the trace metallic impurities is reported on the Certificate of Analysis.
Safety and Handling

Safety and Handling

Causes skin irritation, serious eye irritation. May cause respiratory irritation

DOTInformation : Hazardous

Recommended Storage : Ambient

SDS
Product Certifications

Product intended for laboratory use only