missing translation for 'onlineSavingsMsg'
Learn More

Praseodymium(III) trifluoromethanesulfonate, 98%, Thermo Scientific Chemicals

$192.42 - $192.42

Chemical Identifiers

CAS 52093-27-3
Molecular Formula C3F9O9PrS3
Molecular Weight (g/mol) 588.097
MDL Number MFCD00192417
InChI Key ROUBZIWQWFQCHU-UHFFFAOYSA-K
Synonym praseodymium iii trifluoromethanesulfonate, praseodymium iii triflate, praseodymium trifluoromethanesulfonate, praseodymium 3+ tritriflate, praseodymium iii trifluoromethane-sulfonate, praseodymium iii tris trifluoromethanesulfonate, praseodymium 3+ ion tritrifluoromethanesulfonate
PubChem CID 9894708
IUPAC Name praseodymium(3+);trifluoromethanesulfonate
SMILES C(F)(F)(F)S(=O)(=O)[O-].C(F)(F)(F)S(=O)(=O)[O-].C(F)(F)(F)S(=O)(=O)[O-].[Pr+3]
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AA4031009
View Documents
Thermo Scientific Chemicals
04031009
10 g
Each for $192.42
Only null left
 
Description

Description

It is a water-tolerant Lewis acid used in the Aldol reaction of silyl enol ethers with aldehydes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

52093-27-3
588.097
ROUBZIWQWFQCHU-UHFFFAOYSA-K
9894708
C(F)(F)(F)S(=O)(=O)[O-].C(F)(F)(F)S(=O)(=O)[O-].C(F)(F)(F)S(=O)(=O)[O-].[Pr+3]
C3F9O9PrS3
MFCD00192417
praseodymium iii trifluoromethanesulfonate, praseodymium iii triflate, praseodymium trifluoromethanesulfonate, praseodymium 3+ tritriflate, praseodymium iii trifluoromethane-sulfonate, praseodymium iii tris trifluoromethanesulfonate, praseodymium 3+ ion tritrifluoromethanesulfonate
praseodymium(3+);trifluoromethanesulfonate

Specifications

52093-27-3
10 g
MFCD00192417
Insoluble in water.
C(F)(F)(F)S(=O)(=O)[O-].C(F)(F)(F)S(=O)(=O)[O-].C(F)(F)(F)S(=O)(=O)[O-].[Pr+3]
588.097
588.11
Praseodymium(III) trifluoromethanesulfonate
Powder
C3F9O9PrS3
praseodymium iii trifluoromethanesulfonate, praseodymium iii triflate, praseodymium trifluoromethanesulfonate, praseodymium 3+ tritriflate, praseodymium iii trifluoromethane-sulfonate, praseodymium iii tris trifluoromethanesulfonate, praseodymium 3+ ion tritrifluoromethanesulfonate
ROUBZIWQWFQCHU-UHFFFAOYSA-K
praseodymium(3+);trifluoromethanesulfonate
9894708
98%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only