missing translation for 'onlineSavingsMsg'
Learn More

Praseodymium(III) trifluoromethanesulfonate, 98%, Thermo Scientific Chemicals

Catalog No. AA4031009
Change view
Click to view available options
Quantity:
10 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA4031009 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AA4031009 Supplier Thermo Scientific Chemicals Supplier No. 04031009

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 52093-27-3 | Molecular Formula: C3F9O9PrS3 | Molecular Weight: 588.097 g/mol

It is a water-tolerant Lewis acid used in the Aldol reaction of silyl enol ethers with aldehydes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 52093-27-3
Molecular Formula C3F9O9PrS3
Molecular Weight (g/mol) 588.097
MDL Number MFCD00192417
InChI Key ROUBZIWQWFQCHU-UHFFFAOYSA-K
Synonym praseodymium iii trifluoromethanesulfonate, praseodymium iii triflate, praseodymium trifluoromethanesulfonate, praseodymium 3+ tritriflate, praseodymium iii trifluoromethane-sulfonate, praseodymium iii tris trifluoromethanesulfonate, praseodymium 3+ ion tritrifluoromethanesulfonate
PubChem CID 9894708
IUPAC Name praseodymium(3+);trifluoromethanesulfonate
SMILES C(F)(F)(F)S(=O)(=O)[O-].C(F)(F)(F)S(=O)(=O)[O-].C(F)(F)(F)S(=O)(=O)[O-].[Pr+3]

Specifications

Physical Form Powder
Quantity 10 g
Solubility Information Insoluble in water.
Formula Weight 588.11
Percent Purity 98%
Chemical Name or Material Praseodymium(III) trifluoromethanesulfonate
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only