accessibility menu, dialog, popup

UserName

CAS RN 19423-77-9

ClClClPr

CAS RN 19423-77-9

IUPAC Name: praseodymium(3+) trichloride
Synonyms: praseodymium(III) chloride
Molecular Weight (g/mol): 247.26
Molecular Formula: Cl3Pr
InChi Key: LHBNLZDGIPPZLL-UHFFFAOYSA-K
SMILES: [Cl-].[Cl-].[Cl-].[Pr+3]
Grade 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (1)
  • (1)