missing translation for 'onlineSavingsMsg'
Learn More

Praseodymium AA Standard, 1000 ppm Pr in 3% HNO3, Ricca Chemical

Supplier:  Ricca Chemical Company APR1KN500

Packaging Natural Poly Bottle

 View more versions of this product

Catalog No. APR1KN500

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

Specifications

Chemical Identifiers

12037-29-5, 7697-37-2, 12037-29-5

Specifications

Praseodymium AA Standard
12037-29-5 , 7697-37-2 , 12037-29-5
97.0, 4.53, 0.12
Natural Poly Bottle
94.0, 4.36, 0.11
1mL = 1mg Pr (1, 000ppm Pr)Pr6O in 3% HNO3
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

ShelfLife : 24 months

SDS
Documents

Documents