missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ PRAK Monoclonal Antibody (7H10B4)

Description
MA5-15352 targets PRAK in indirect ELISA, IHC and WB applications and shows reactivity with Human samples. The MA5-15352 immunogen is purified recombinant fragment of PRAK expressed in E. Coli.
MAPKAPK5 (PRAK) is activated in response to cellular stress and pro-inflammatory cytokines. Activity of this kinase is regulated by p38 alpha and p38 beta. The protein encoded by PRAK is a member of the serine/threonine kinase family. In response to cellular stress and proinflammatory cytokines, this kinase is activated through its phosphorylation by MAP kinases including MAPK1/ERK, MAPK14/p38-alpha, and MAPK11/p38-beta. In vitro, this kinase phosphorylates heat shock protein HSP27 at its physiologically relevant sites. Two alternately spliced transcript variants of PRAK encoding distinct isoforms have been reported.
Specifications
Specifications
| Antigen | PRAK |
| Applications | ELISA, Immunohistochemistry (Paraffin), Western Blot |
| Classification | Monoclonal |
| Clone | 7H10B4 |
| Concentration | Conc. Not Determined |
| Conjugate | Unconjugated |
| Formulation | Ascites with 0.03% sodium azide |
| Gene | MAPKAPK5 |
| Gene Accession No. | Q8IW41 |
| Gene Alias | MAP kinase-activated protein kinase 5; MAPK-activated protein kinase 5; MAPKAP kinase 5; Mapkapk5; MAPKAP-K5; MAPKAPK-5; mitogen-activated protein kinase-activated protein kinase 5; MK5; MK-5; p38-regulated/activated protein kinase; PRAK |
| Show More |