missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PPP1R9B Monoclonal Antibody (K94019_12A3), Invitrogen™
Mouse Monoclonal Antibody
$1421.98 - $2134.69
Specifications
| Antigen | PPP1R9B |
|---|---|
| Clone | K94019_12A3 |
| Concentration | 1 mg/mL |
| Applications | Western Blot |
| Classification | Monoclonal |
Description
Sequence of this protein is as follows: FSTAPIQVFS TYSNEDYDRR NEDVDPMAAS AEYELEKRVE RLELFPVELE KDSEGLGISI IGMGAGADMG LEKLGIFVKT VTEGGAAHRD GRIQVNDLLV EVDGTSLVGV TQSFAASVLR NTKGRVRFMI GRERPGEQSE VAQLIQQTLE QERWQREMME QRYAQYGEDD EETGEYATDE DEELSPTFPG GEMAIEVFEL AENEDALSPV DMEPEKLVHK FKELQIKHAV TEAEIQQLKR KLQSLEQEKG RWRVEKAQLE QSVEENKERM EKLEGYWGEA QSLCQAVDEH LRETQAQYQA LERKYSKAKR LIKDYQQKEI EFLKKETAQR RVLEESELAR KEEMDKLLDK ISELEGNLQT
PPP1R9B Monoclonal antibody specifically detects PPP1R9B in Human, Rat samples. It is validated for Western BlotSpecifications
| PPP1R9B | |
| 1 mg/mL | |
| Monoclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol, 0.5% BSA and 0.09% sodium azide | |
| Neb2; neurabin 2; neurabin II; neurabin-2; Neurabin-II; neural tissue-specific F-actin-binding protein II; nuerabin 2; p130; PP1bp134; PPP1R6; PPP1R9; PPP1R9B; Protein phosphatase 1 regulatory subunit 9B; protein phosphatase 1, regulatory (inhibitor) subunit 9B; protein phosphatase 1, regulatory subunit 9B; protein phosphatase 1, regulatory subunit 9B, spinophilin; SPINO; Spinophilin; SPL; Spn | |
| Recombinant human PPP1R9B (immunogen range is 451-810). | |
| Primary | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Ppp1r9b |
| K94019_12A3 | |
| Western Blot | |
| Unconjugated | |
| Mouse | |
| Human, Rat | |
| O35274, Q96SB3 | |
| 84686, 84687 | |
| IgG1 | |
| Protein A | |
| Antibody |