missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ PPP1R3D Polyclonal Antibody

Description
Antibody detects endogenous levels of total PPP1R3D.
Seems to act as a glycogen-targeting subunit for PP1. PP1 is essential for cell division, and participates in the regulation of glycogen metabolism, muscle contractility and protein synthesis.
Specifications
Specifications
| Antigen | PPP1R3D |
| Applications | Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 |
| Gene | PPP1R3D |
| Gene Accession No. | O95685 |
| Gene Alias | 2610024M03Rik; Gm359; PP1 subunit R6; PPP1R3D; PPP1R6; protein phosphatase 1 regulatory subunit 3D; Protein phosphatase 1 regulatory subunit 6; protein phosphatase 1, regulatory (inhibitor) subunit 3D; protein phosphatase 1, regulatory subunit 3D; protein phosphatase 1, regulatory subunit 6, spinophilin; protein phosphatase 1-binding subunit R6 |
| Gene Symbols | PPP1R3D |
| Show More |