missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ PPP1R16A Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA521661
Description
Recommended positive controls: 293T, A431, H1299, HeLaS3, HepG2, Molt-4, Raji, mouse liver. Predicted reactivity: Mouse (84%), Rat (85%), Rhesus Monkey (97%), Bovine (87%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
Inhibits protein phosphatase 1 activity toward phosphorylase, myosin light chain and myosin substrates. Jikuya H., Submitted (JUL-2002) to the EMBL/GenBank/DDBJ databases. Kalnine N., Submitted (MAY-2003) to the EMBL/GenBank/DDBJ databases. The MGC Project Team, Genome Res. 14:2121-2127(2004).Chemical Identifiers
| PPP1R16A | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Polyclonal | |
| Liquid | |
| IgG | |
| Human, Mouse | |
| PPP1R16A | |
| 2900084E10Rik; mFLJ00392 protein; myosin phosphatase targeting subunit 3; myosin phosphatase, target subunit 3; myosin phosphatase-targeting subunit 3; Mypt3; Ppp1r16a; Protein phosphatase 1 regulatory subunit 16A; protein phosphatase 1, regulatory (inhibitor) subunit 16A; protein phosphatase 1, regulatory subunit 16A; R75527 | |
| PPP1R16A | |
| Primary | |
| Antigen affinity chromatography |
| 0.86 mg/mL | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| RUO | |
| 0.1M tris glycine with 10% glycerol and 0.01% thimerosal; pH 7 | |
| Q923M0, Q96I34 | |
| 73062, 84988 | |
| Recombinant protein encompassing a sequence within the center region of human PPP1R16A. The exact sequence is proprietary. | |
| Antibody |
Specifications
| PPP1R16A | |
| Polyclonal | |
| Unconjugated | |
| PPP1R16A | |
| 2900084E10Rik; mFLJ00392 protein; myosin phosphatase targeting subunit 3; myosin phosphatase, target subunit 3; myosin phosphatase-targeting subunit 3; Mypt3; Ppp1r16a; Protein phosphatase 1 regulatory subunit 16A; protein phosphatase 1, regulatory (inhibitor) subunit 16A; protein phosphatase 1, regulatory subunit 16A; R75527 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 73062, 84988 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot | |
| 0.86 mg/mL | |
| 0.1M tris glycine with 10% glycerol and 0.01% thimerosal; pH 7 | |
| Q923M0, Q96I34 | |
| PPP1R16A | |
| Recombinant protein encompassing a sequence within the center region of human PPP1R16A. The exact sequence is proprietary. | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |