missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ PPM1G Recombinant Rabbit Monoclonal Antibody (23GB2585)
GREENER_CHOICE

Catalog No. MA553540
Change view
Click to view available options
Quantity:
20 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MA553540 100 μL
MA553541 20 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MA553540 Supplier Invitrogen™ Supplier No. MA553540
Only null left

Rabbit Recombinant Monoclonal Antibody

PPM1G Recombinant Monoclonal Antibody for Western Blot, ICC/IF, Flow

The protein encoded by this gene is a member of the PP2C family of Ser/Thr protein phosphatases. PP2C family members are known to be negative regulators of cell stress response pathways. This phosphatase is found to be responsible for the dephosphorylation of Pre-mRNA splicing factors, which is important for the formation of functional spliceosome. Studies of a similar gene in mice suggested a role of this phosphatase in regulating cell cycle progression. Alternatively spliced transcript variants encoding the same protein have been described.

Specifications

Antigen PPM1G
Applications Flow Cytometry, Western Blot, Immunocytochemistry
Classification Recombinant Monoclonal
Clone 23GB2585
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene Ppm1g
Gene Accession No. F1LNI5, O15355, Q61074
Gene Alias AI385652; fibroblast growth factor inducible 13; fibroblast growth factor-inducible protein 13; Fin13; PP2C, gamma; PP2CG; PP2CGAMMA; PP2C-gamma; Ppm1c; Ppm1g; PPP2CG; protein phosphatase 1C; Protein phosphatase 1G; protein phosphatase 1G (formerly 2C), magnesium-dependent, gamma isoform; protein phosphatase 2, catalytic subunit, gamma isoform; protein phosphatase 2C gamma isoform; protein phosphatase 2C isoform gamma; Protein phosphatase magnesium-dependent 1 gamma; protein phosphatase, Mg2+/Mn2+ dependent 1G; protein phosphatase, Mg2+/Mn2+ dependent, 1G
Gene Symbols Ppm1g
Host Species Rabbit
Immunogen A synthesized peptide derived from human PPM1G (301-450AA).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 14208, 259229, 5496
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less