missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PPM1G Rabbit anti-Human, Mouse, Polyclonal, Bethyl Laboratories
Description
The recommended shelf life for this product is 1 year from date of receipt.
The protein encoded by this gene is a member of the PP2C family of Ser/Thr protein phosphatases. PP2C family members are known to be negative regulators of cell stress response pathways. This phosphatase is found to be responsible for the dephosphorylation of Pre-mRNA splicing factors, which is important for the formation of functional spliceosome. Studies of a similar gene in mice suggested a role of this phosphatase in regulating cell cycle progression. Alternatively spliced transcript variants encoding the same protein have been described.
Specifications
Specifications
| Antigen | PPM1G |
| Applications | Immunoprecipitation, Western Blot |
| Classification | Polyclonal |
| Concentration | 0.2 mg/ml |
| Conjugate | Unconjugated |
| Formulation | TBS with 0.1% BSA and 0.09% sodium azide |
| Gene | PPM1G |
| Gene Accession No. | O15355, Q61074 |
| Gene Alias | Protein phosphatase 2C gamma isoform; PP2C-gamma; Protein phosphatase magnesium-dependent 1 gamma; Protein phosphatase 1C; PPM1C |
| Gene Symbols | PPM1G |
| Show More |