missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ PPFIA4 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA562787
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA562787 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA562787 Supplier Invitrogen™ Supplier No. PA562787
Season of Savings
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Rabbit Polyclonal Antibody

Immunogen sequence: ERVTTLEEQL AGAHQQVSAL QQGAGVRDGA AEEEGTVELG PKRLWKEDTG RVEELQ Highest antigen sequence identity to the following orthologs: Mouse - 93%, Rat - 91%.

PPFIA4, or liprin-alpha-4, belongs to the liprin-alpha gene family. See liprin-alpha-1 (LIP1, or PPFIA1; MIM 611054) for background on liprins.

Specifications

Antigen PPFIA4
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.30 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene PPFIA4
Gene Accession No. O75335
Gene Alias 1110008G13Rik; AI448359; AI852265; C81506; KIAA0897; Liprin-alpha4; Liprin-alpha-4; LOW QUALITY PROTEIN: liprin-alpha-4; PPFIA4; Protein tyrosine phosphatase receptor type f polypeptide-interacting protein alpha-4; protein tyrosine phosphatase, receptor type, f polypeptide (PTPRF), interacting protein (liprin), alpha 4; PTPRF interacting protein alpha 4; PTPRF-interacting protein alpha-4
Gene Symbols PPFIA4
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human PPFIA4. Recombinant protein control fragment (Product #RP-100956).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 8497
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less