missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ PP1 gamma Polyclonal Antibody
GREENER_CHOICE

Catalog No. 01674114
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01674114 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01674114 Supplier Invitrogen™ Supplier No. PA112381
Only null left

Rabbit Polyclonal Antibody

This antibody will not cross-react with other PP1 isoforms.

The protein encoded by this gene belongs to the protein phosphatase family, PP1 subfamily. PP1 is an ubiquitous serine/threonine phosphatase that regulates many cellular processes, including cell division. It is expressed in mammalian cells as three closely related isoforms, alpha, beta/delta and gamma, which have distinct localization patterns. This gene encodes the gamma isozyme. Alternatively spliced transcript variants encoding different isoforms have been found for this gene.

Specifications

Antigen PP1 gamma
Applications Immunoprecipitation, Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.08% sodium azide
Gene PPP1CC
Gene Accession No. P36873, P61287, P63087, P63088
Gene Alias dis2m1; MGC1675; MGC2870; PP1; PP1C; PP1C gamma 1; PP1C gamma 2; PP1G; PP-1G; PP1gamma; PP2CG; PP2CGAMMA; PPP1CC; Ppp1cc1; PPP1G; PPP2CG; protein phosphatase 1 catalytic subunit gamma; protein phosphatase 1, catalytic subunit, gamma isoform; protein phosphatase 1, catalytic subunit, gamma isozyme; protein phosphatase 1C catalytic subunit; protein serine-threonine phosphatase catalytic subunit PP-1b; serine/threonine phosphatase 1 gamma; serine/threonine-protein phosphatase PP1-gamma catalytic subunit
Gene Symbols PPP1CC
Host Species Rabbit
Immunogen Synthetic peptide corresponding to C-termianl region of PP1 gamma 1 catalytic subunit (amino acids 311-323).
Purification Method Ammonium sulfate precipitation
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 19047, 24669, 282318, 5501
Target Species Human, Mouse, Rat, Bovine
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less