missing translation for 'onlineSavingsMsg'
Learn More

SW Sustainability Solutions PowerChem™ PC-12WT100 EcoTek Sustainable Nitrile Cleanroom Gloves

Catalog No. 18139435
Change view
Click to view available options
Size:
Small
Medium
Large
X-Large
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
18139435 X-Large
18139432 Small
18139433 Medium
18139434 Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 18139435 Supplier SW Sustainability Solutions Supplier No. N427105

High-performance nitrile cleanroom gloves deliver exceptional durability, chemical resistance, and particulate control for controlled environments.

  • EcoTek™ Sustainable Technology
  • SafeDon™ dispensing minimizes cross-contamination while delivering significant space and packaging savings. (Save over 40% on space and storage and reduce cardboard waste by 20% compared to conventional 100-glove boxes.)
  • Low Dermatitis Potential (LDP) minimizes the risk of Type I and Type IV allergies and enhances safety for users with sensitive skin
  • Class 100 (ISO 5) cleanroom gloves
  • Extended-cuff for additional protection
  • Optimal thickness for durability and tactile sensitivity
  • Tear-resistant beaded cuff eases donning
  • Finger textured surface delivers excellent grip
  • Extended-cuff for added coverage and protection
  • SHA Dermatologically Approved
  • 1.5 AQL for wet and dry environments

Specifications

Chlorinated Yes
Cleanroom Class Class 100 (ISO 5)
Accelerator Free No
AQL 1.5
Color White
Disposable Single-use
Exterior Finish Fingertip Textured
Finger Thickness (English) 6.2 mil
Finger Thickness (Metric) 0.15 mm
Hand Ambidextrous
Latex Free Yes
Material Nitrile
Palm Thickness (English) 5.0 mil
Palm Thickness (Metric) 0.12 mm
Powdered Powder Free
Product Type Controlled Environment Gloves
Size X-Large
Sterility Non-sterile
Thickness (English) 5.0 mil
Thickness (Metric) 0.12 mm
Model N427105
Packaging Type Cleanroom Bagged
Series PC-12WT
Show More Show Less

EcoTek gloves tested to ASTM D5526-12 Standard Test Method for Determining Anaerobic Biodegradation of Plastic Materials Under Accelerated Landfill Conditions. Results are 92.6% biodegradation in 60% solids landfill at 945 days. Future results cannot be predicted/extrapolated. If you need more information on the test reports, send email to cs@swssglobal.com.