missing translation for 'onlineSavingsMsg'
Learn More

Power Blotter Welcome Pack, PVDF

Catalog No. PB0116
Change view
Click to view available options
Quantity:
1 Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PB0116 1 Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PB0116 Supplier Thermo Fisher Scientific Supplier No. PB0116

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

The Invitrogen Power Blotter System (Power Blotter Station + Power Blotter Cassette) is designed specifically for rapid semi-dry transfer of proteins from polyacrylamide gels to nitrocellulose or PVDF membranes in 5 to 12 minutes.

The Power Blotter System features an integrated power supply optimized to enable consistent and high-efficiency protein transfer.

The Power Blotter Welcome Pack, PVDF, includes:

  • Power Blotter Station
  • Power Blotter Cassette
  • Power Blotter Select Transfer Stacks, PVDF, regular size
  • Power Blotter Pre-cut Membranes and Filters, PVDF, regular size
  • Power Blotter 1-Step Transfer Buffer

Features of the Power Blotter System include:

  • Efficient—high transfer efficiency with a broad range of protein sizes compared to conventional semi-dry or wet (tank) transfer methods
  • Fast—5- to 12-minute transfer when used with Power Blotter Select Transfer Stacks or Power Blotter 1-Step Transfer Buffer
  • Integrated power supply—seamless operation between control unit and cassette; no need for external power supply
  • Easy-touch programming—convenient pre-programmed transfer methods, or create, save, and run customized transfer methods
  • Flexible—simultaneous transfer of 1 or 2 mini-size or 1 midi-size gel using the Power Blotter Cassette
  • Versatile—compatible with Power Blotter Select Transfer Stacks and Pierce 1-Step Transfer Buffer for rapid blotting programs or Towbin transfer buffer for conventional semi-dry transfer methods

The Power Blotter System achieves highly efficient transfer in seven minutes when used with Power Blotter Select Transfer Stacks or ten minutes with Power Blotter 1-Step Transfer Buffer. The instrument is also effective for standard 30- to 60-minute semi-dry transfer protocols based on traditional buffers.

Specifications

For Use With (Equipment) Acrylamide Gels
Quantity 1 Kit
Material PVDF
Content And Storage • Power Blotter Station
• Power Blotter Cassette
• Power Blotter Select Transfer Stacks, PVDF, regular-size
• Power Blotter Pre-cut Membranes and Filters, PVDF, regular-size
• Power Blotter 1-Step Transfer Buffer

Shipping Condition Room Temperature
Mode of Transfer Semi-Dry
Product Line Power Blotter

For Research Use Only. Not for use in diagnostic procedures.