missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Povidone K-30, USP, Spectrum™ Chemical
P1454, 9003-39-8, (C6H9NO)n
Supplier: Spectrum Chemical Mfg Cor P145412KGBL
Description
Spectrum™ Chemical Povidone K-30, USP is used as a binder in pharmaceutical tablets. All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.Specifications
| 9003-39-8 | |
| 3.0 to 5.0 | |
| Fiber Drum | |
| MFCD01076626 | |
| WHNWPMSKXPGLAX-UHFFFAOYSA-N | |
| 1-ethenylpyrrolidin-2-one | |
| USP |
| 1 | |
| 0.001 | |
| (C6H9NO)n | |
| 12 kg | |
| *-CC(-*)N1CCCC1=O | |
| 111.14 |
Chemical Identifiers
| 9003-39-8 | |
| 111.14 | |
| WHNWPMSKXPGLAX-UHFFFAOYSA-N | |
| *-CC(-*)N1CCCC1=O |
| (C6H9NO)n | |
| MFCD01076626 | |
| 1-ethenylpyrrolidin-2-one |