missing translation for 'onlineSavingsMsg'
Learn More

Povidone K-30, USP, Spectrum™ Chemical

Catalog No. p-7158080
Change view
Click to view available options
Quantity:
100 g
5 kg
12 kg
50 kg
Packaging:
Fiber Drum
Poly Bottle
Poly Pail
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18605747 100 g Poly Bottle
18605749 5 kg Poly Pail
18605750 12 kg Fiber Drum
18605751 50 kg Fiber Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 18605747 Supplier Spectrum Chemical Mfg Cor Supplier No. P1454100GM
Only null left

P1454, 9003-39-8, (C6H9NO)n

Spectrum™ Chemical Povidone K-30, USP is used as a binder in pharmaceutical tablets. All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 9003-39-8
Molecular Formula (C6H9NO)n
Molecular Weight (g/mol) 111.14
MDL Number MFCD01076626
InChI Key WHNWPMSKXPGLAX-UHFFFAOYSA-N
IUPAC Name 1-ethenylpyrrolidin-2-one
SMILES *-CC(-*)N1CCCC1=O

Specifications

CAS Max % 1
pH 3.0 to 5.0
Ignition Residue 0.001
Packaging Poly Bottle
Quantity 100 g
Grade USP